SYNF608350F
Synthware™ Buchner funnel
350 mL, porosity: fine
Synonym(e):
Synthware F608350F, filtering funnel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F608350F
Kapazität
350 mL
Verwandte Kategorien
Allgemeine Beschreibung
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)