SYNF459125A
Synthware™ Squibb-style separatory funnel with standard taper joints, PTFE stopcock and PTFE stopper
125 mL, joint: ST/NS 19/22, PTFE stopcock and glass stopper
Synonym(e):
Synthware F459125A, glass separatory funnel
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F459125A
Kapazität
125 mL
125 mL
Allgemeine Beschreibung
With 2 mm PTFE stopcock. Supplied with a glass stopper or PTFE stopper.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)