Merck
SYNF410250
Synthware two neck round bottom flask with angled side neck
250 mL, center joint: ST/NS 19/22, side joint: ST/NS 14/20
Synonym(e):
Synthware F410250, 2-neck round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
two-neck
Verbindung
center joint: ST/NS19/22
side joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F410250
Kapazität
250 mL
250 mL
Allgemeine Beschreibung
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)