Merck
SYNF271018M
Synthware fritted gas dispersion tube with 15 degree angle
disc diam. 10 mm, porosity: medium, stem O.D. × L 8 mm × 150 mm
Synonym(e):
Synthware F271018M, glass gas dispersion tube
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Materialien
colorless tube
glass tube
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F271018M
Scheibendurchm.
10 mm
Schaft AD × L
8 mm × 150 mm
Allgemeine Beschreibung
The 15° angle permits tube to be inserted into vessels where a large opening is not available. Stem is 8 mm O.D. × 150 mm length, XC stands for extra coarse porosity.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)