Merck
SYNF21842L
Synthware four-neck round-bottom flask with vertical side necks
capacity 2,000 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Synonym(e):
Synthware F21842L, 4-neck reaction flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
four-neck
round bottom
Verbindung
center joint: ST/NS29/42
side joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F21842L
Kapazität
2,000 mL
Allgemeine Beschreibung
With identical vertical side joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)