Merck
SYNF124500
Synthware single-neck flat-bottom flask
capacity 500 mL, joint: ST/NS 24/40
Synonym(e):
Synthware F124500, glass round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
flat bottom
single-neck
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F124500
Kapazität
500 mL
Allgemeine Beschreibung
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)