SYND240001
Synthware™ distillation column packing, mini coil style
O.D. 4 mm, L 12 mm, weight 500 g
Synonym(e):
Synthware D240001
Alle Fotos(1)
About This Item
UNSPSC-Code:
40161600
NACRES:
NB.51
Empfohlene Produkte
Hersteller/Markenname
D240001
Länge
12 mm
AD
4 mm
Gewicht
500 g
Allgemeine Beschreibung
Synthware™ distilling column packing material is a particle-type mini-glass coil packing that can be "poured" into any laboratory or pilot-scale column. It is easily removable for replacement or cleaning.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)