Merck
SYND211150
Synthware vacuum jacketd Vigreux distillation head
joint: ST/NS 14/20, column H 150 mm, immersion depth 25 mm
Synonym(e):
Synthware D211150, short path head, Vigreux distilling head
About This Item
UNSPSC-Code:
41104812
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware D211150
Säule H
150 mm
Eintauchtiefe
25 mm
Verwandte Kategorien
Allgemeine Beschreibung
Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. Vacuum adapter has an inner joint and a sealed in drip tip. To be used with a 25 mm immersion thermometer
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)