SYNC561100
Synthware™ coil-style reflux condenser with removable hose connections
joint: ST/NS 14/20, hose size 8 mm
Synonym(e):
Synthware C561100, coil-type condenser
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C561100
Schlauchgröße
8 mm
Verwandte Kategorien
Allgemeine Beschreibung
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)