Merck
SYNA881420G
Synthware vacuum/inert gas adapter with T-bore glass stopcock
joint: ST/NS 14/20
Synonym(e):
Synthware A881420G, T-shape vacuum gas adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A881420G
Verwandte Kategorien
Allgemeine Beschreibung
With 4 mm T-bore glass stopcock having constant pressure retention device or 2 mm T-bore 1:5 taper PTFE plug.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)