SYNA792434
Synthware™ bushing adapter
female joint: ST/NS 24/40, male joint: ST/NS 34/45
Synonym(e):
Synthware A792434, glass bushing adapter
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
female joint: ST/NS24/40
male joint: ST/NS34/45
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A792434
Allgemeine Beschreibung
Bushing adapters allow for compact assemblies of dissimilar joints.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNA792434-1EA | 4061832189796 |