Merck
SYNA551420
Synthware distillation distribution adapter
joint: ST/NS 14/20
Synonym(e):
Synthware A551420, cow distillation adapter
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A551420
Allgemeine Beschreibung
Rotating cow receiver allows four flasks to be progressively filled.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)