Merck
SYNA272250
Synthware anti-climb adapter
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
Synonym(e):
Synthware A272250, rotary evaporator adapter
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Verbindung
bottom joint: ST/NS14/20
joint: ST/NS top24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A272250
Kapazität
250 mL
Verwandte Kategorien
Allgemeine Beschreibung
Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the The inner vapor tube is sealed to the top of thetrap to prevent entry into the condenser.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)