Merck
SLW1517/10
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 1000 mL
Synonym(e):
Pyrex, SciLabware 1517/10
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Kapazität:
1000 mL
Durchm. × H:
101 mm × 225 mm
Materialien:
borosilicate glass bottle
round bottle
white graduation
Leistungsmerkmale:
graduations
without cap
with printed trace code
without pouring ring
Verpackung:
pack of 10 ea
Materialien
borosilicate glass bottle
round bottle
white graduation
Verbindung
Joints threaded neck (GL45)
Leistungsmerkmale
graduations
without cap
with printed trace code
without pouring ring
Verpackung
pack of 10 ea
Hersteller/Markenname
SciLabware 1517/10
Kapazität
1000 mL
Durchm. × H
101 mm × 225 mm
Allgemeine Beschreibung
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Leistungsmerkmale und Vorteile
• Bottles only. Supplied without cap or pouring ring.• Complies with ISO 4796.• Printed with Trace Code for downloadable batch certificate.• Sloping shoulders for smooth pouring action.• White graduations and marking spot.• Manufactured from temperature and chemically resistant borosilicate glass.• Choice of caps and pouring rings available separately.• The 25 ml size bottle features a glass lip and does not require a pouring ring.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)