Merck
PL800GH-8
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
Synonym(e):
Plas-Labs Glove Box
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Materialien
Hypalon®
Größe
8 (for 8 in. ports)
Leistungsmerkmale
reusable
Verpackung
pack of 1 pair
Hersteller/Markenname
Plas-Labs 800GH-8
Allgemeine Beschreibung
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Rechtliche Hinweise
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
PL800GH-8-1PR4061832875743