Merck
CLS882114
Pyrex® thermometer adapter
joint: ST/NS 14/20
Synonym(e):
lab equipment accessory
About This Item
UNSPSC-Code:
41121815
Verbindung
joint: ST/NS14/20
Hersteller/Markenname
Corning 8821-14
Länge
60 mm
Allgemeine Beschreibung
One end with a ground joint and the other end tooled to accommodate a thermometer.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)