CLS6404250
Pyrex® Squibb separatory funnel, PE stopper, PTFE stopcock
capacity 250 mL
Synonym(e):
Corning® 6404
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Empfohlene Produkte
Verbindung
joint stopper22
Hersteller/Markenname
Corning 6404-250
Kapazität
250 mL
Größe
Absperrhahngröße
4
Verwandte Kategorien
Allgemeine Beschreibung
PYREX® pear-shaped glass separatory funnels contain stoppers and polytetrafluorethylene (PTFE) stopcocks. The closed bottom stopper made of linear high density polyethylene to conform to standard taper stopper dimensions. The PTFE plug is designed to fit into the micro-finish barrel and it also minimizes freezing problems and grease contamination.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)