Merck
CLS61500500
PYREXPLUS® narrow-mouth reagent bottle, with Pyrex® ST stopper, protective coating
capacity 500 mL
Synonym(e):
Corning® 61500
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle
polypropylene cap
Stopfengröße
24
Leistungsmerkmale
autoclavable
cap
Verpackung
pack of 1 ea
case of 6 ea
Hersteller/Markenname
Corning 61500-500
Flaschenhalsdurchm.
~35.5 mm
Kapazität
500 mL
Durchm. × H
86 mm × 163.5 mm
Fitting
45
Allgemeine Beschreibung
Features a protective plastic coating that helps prevent glass from shattering and reduces any spills. Fully autoclavable (121°C) and resistant to thermal shock. Bottle has a ground neck,
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
CLS61500500-6EA4061831675153
CLS61500500-1EA