Merck
CLS563110
Pyrex® certified and serialized micro volumetric flask, with Pyrex® stopper
capacity 10 mL
Synonym(e):
flasks glass, volumetric flasks, volumetric glass, Corning® 5631
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
long
Stopfengröße
9
Sterilität
non-sterile
Leistungsmerkmale
closure type ST stopper
Verpackung
case of 6
Hersteller/Markenname
Corning 5631-10
tol. measuring range
0.02 mL
Kapazität
10 mL
Allgemeine Beschreibung
Calibrated to Class A tolerance and individually serialized and supplied with at Certificate of Identification and Capacity. Hexagonal base.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)