CLS4980PACK
Pyrex® narrow-mouth graduated Erlenmeyer flask assortment pack
assortment pack
Synonym(e):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks glass
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass
narrow-mouth
Sterilität
non-sterile
Leistungsmerkmale
cap: no
Verpackung
case of 1
Hersteller/Markenname
Corning 4980-PACK
Fitting
cap for GL 45
Allgemeine Beschreibung
A convenience pack containing one each of the most popular sizes of Cat. No. 4980 flask. Designed for the low volume user, a case contains one each of five sizes: 50 mL, 125 mL, 250 mL, 500 mL, and 1 L.
Sonstige Hinweise
Browse all Erlenmeyer flasks
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)