CAD6011
3-way stopcock
female Luer lock to male Luer lock, right side, female Luer lock, compression-nut
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122000
NACRES:
NB.03
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Cadence Science 6011
Verwandte Kategorien
Allgemeine Beschreibung
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
- Made from nickel plated brass or stainless steel
- Tested to 45 psi water pressure
- High pressure stopcocks are rated to 290 psi
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6011-1EA | 4061832517223 |