BR91263
BRAND® glass beaker with spout, low form
volume 2000 mL, with graduation
Synonym(e):
Beakers, Glass
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass 3.3
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 91263
Volumen
2000 mL
Allgemeine Beschreibung
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)