BR87610
BRAND® PP beaker with spout, low form
volume 150 mL
Synonym(e):
Beakers, Plastics
Alle Fotos(3)
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene
Beschreibung
in addition to ISO 7056
Leistungsmerkmale
graduations
beaded rim and spout
Hersteller/Markenname
BRAND 87610
Volumen
150 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)