BR82170
BRAND® needle-valve stopcocks for burettes and automatic burettes with PTFE spindle
Straight, for burette capacity 2-50 mL
Synonym(e):
Accessories, Burettes, Needle-valve stopcock, Stopcocks
Alle Fotos(3)
About This Item
UNSPSC-Code:
40141602
NACRES:
NB.43
Empfohlene Produkte
Materialien
polystyrene
Leistungsmerkmale
for burette capacity 2-50 mL
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 82170
Allgemeine Beschreibung
Interchangeable PTFE valve spindle, PP cap and retention device. Valve opening 0-25.5 mm.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Please note: PTFE valve spindle operates smoothly yet is tight, even without lubrication, so there is no grease residue on the burette. The valve spindle is replaceable. Tight seal without force, a gentle turn is all it takes to close the burette valve.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)