BR66015
BRAND® dessicator plate
for nominal size 150 mm, O.D. 140 mm, polypropylene
Synonym(e):
Accessories, Desiccator plate, Desiccators
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 66015
AD
140 mm
Eignung
suitable for (glass dessicators)
Verwandte Kategorien
Allgemeine Beschreibung
Our porcelain plates for glass desiccators and polypropylene (PP) plates for PC/PP desiccators are essential components designed to enhance the functionality and efficiency of your desiccation processes. Both porcelain and PP plates are designed to optimize the performance of your desiccators, ensuring that your samples remain protected from moisture and environmental contaminants.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR66015-1EA | 4061831866308 |