BR474701
BRAND® microscope slide
cut edges
Synonym(e):
Microscope slides, Microscopy
Alle Fotos(2)
About This Item
UNSPSC-Code:
41122600
NACRES:
MA.01
Empfohlene Produkte
Materialien
soda-lime glass
Leistungsmerkmale
cut edges
Hersteller/Markenname
BRAND 474701
Größe
76 mm × 26 mm × 1 mm
Allgemeine Beschreibung
- Made from clear soda-lime glass, hydrolytic class 3.
- Ground edges are recommended to reduce the risk of injury.
- Thickness approx.: 1 mm
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)