BR36408
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 50 mL, polypropylene stopper, joint: ST/NS 12/21
Synonym(e):
BLAUBRAND®, Volumetric flasks
About This Item
Empfohlene Produkte
Materialien
polypropylene stopper
transparent borosilicate glass 3.3 flask
Verbindung
joint: ST/NS12/21
Leistungsmerkmale
accuracy class A (To contain)
Verpackung
pack of 2 ea
Hersteller/Markenname
BRAND 36408
Kapazität
50 mL
Verwandte Kategorien
Allgemeine Beschreibung
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR36408-2EA | 4061831865301 |