BR25900
BRAND® pipette controller micro-classic
Synonym(e):
Pipette Controller, Pipetting Aids, micro Pipette Controller
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Materialien
(Plastic & metal with autoclavable tube)
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 25900
Volumen
1 mL
Verwandte Kategorien
Allgemeine Beschreibung
Working under a microscope requires utmost concentration and therefore comfortable and reliable instruments. The micro-classic pipette controller with its ergonomic shape and simple handling offers comfort and convenience for this strenuous job. It is a must in IVF and medical laboratories. Suitable for disposable micropipettes with ring mark and other small volume pipettes up to 1 mL (e.g., blood diluting pipettes). The micro-classic adapts to right- and left-handed operation. Adapter and suction tube are autoclavable at 121 °C. The micro-classic minimizes the risk of contamination when working with infectious material. includes 2 spare suction tubes.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
BR25900-1EA | 4061831864861 |