BR152510
BRAND® tubing adapter
high-density polyethylene, for tubing i.d., 12-14 to 18-20,5 mm
Synonym(e):
Tubing, Tubing adapters
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.43
Empfohlene Produkte
Materialien
high-density polyethylene
Leistungsmerkmale
for tubing i.d. 12-14 to 18-20,5 mm
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 152510
Länge
73 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR152510-10EA | 4061831854176 |