BR136325
BRAND® stirring rod
high-density polyethylene (with steel core), L × D 300 mm × 6 mm
Synonym(e):
PTFE, Stirring rods
Alle Fotos(3)
About This Item
UNSPSC-Code:
41121802
NACRES:
NB.72
Empfohlene Produkte
Materialien
high-density polyethylene (with steel core)
Beschreibung
with steel core
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 136325
L × T
300 mm × 6 mm
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® stirring rod is designed to be bendable, allowing for customization based on specific applications. The permanent bend ensures that the rod maintains its shape during use, providing flexibility for stirring in various vessel shapes and sizes. This innovative design enhances the efficiency of mixing and blending processes, making it suitable for a wide range of laboratory tasks.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)