BR130324
BRAND® screw cap
thread for GL W 54
Synonym(e):
Accessories, Bottles, Screw caps
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Materialien
polypropylene
Beschreibung
for wide neck bottles
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 130324
Fitting
thread for GL W 54
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Screw Cap is a secure blue cap designed for square bottles, featuring a tamper-proof closure that ensures the integrity of the contents. This cap includes a press-off retaining ring and a sealing cone, providing an effective seal to prevent leaks and contamination. Its durable construction makes it suitable for various laboratory and industrial applications, ensuring reliable performance in sample storage and handling.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)