BR122710
BRAND® Media Bottle
capacity 500 mL, borosilicate glass 3.3 bottle, polypropylene cap, pack of 4 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Materialien
borosilicate glass 3.3 bottle
polypropylene cap
round bottle
Verbindung
Joints threaded neck (GL56)
Sterilität
non-sterile
Leistungsmerkmale
graduations
with cap
Verpackung
pack of 4 ea
Hersteller/Markenname
BRAND 122710
Kapazität
500 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Media Bottle is expertly crafted from Boro 3.3 glass, ensuring excellent durability and resistance to thermal and chemical stress. Designed with a GL 56 neck, this bottle features a low height for easy handling and storage. The PP screw-on cap provides a secure, vacuum-tight seal, making it ideal for sterile filtration applications. The bottle is also equipped with a clear scale for accurate volume measurement, enhancing its usability in laboratory settings. This versatile media bottle is essential for storing and handling various media and solutions, ensuring reliability and safety in your experiments.
Leistungsmerkmale und Vorteile
- Bottle and cap made of biologically inert material
- Larger bottle neck and bottle can be tilted by 45° for easy removal of medium
- Compatible with bottle top filters
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR122710-4EA | 4061835293858 |