Merck
BAH131780612
SP Bel-Art® utility bags
width × H × thickness 6 in. (152 mm) × 12 in. (305 mm) × 4 mil (0.1 mm), clear, pack of 100 ea
Synonym(e):
clear bag, reusable bag, sample bag
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
clear
Verpackung
pack of 100 ea
Hersteller/Markenname
Bel-Art H13178-0612
width × H × thickness
6 in. (152 mm) × 12 in. (305 mm) × 4 mil (0.1 mm)
Allgemeine Beschreibung
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Leistungsmerkmale und Vorteile
  • FDA grade polyethylene, safe for food, drug and cosmetic use
  • Latex free
  • Not Autoclavable
  • H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)