BAF371190083
Spinbar® Magnetrührstäbchen
white, L × diam. 0.315 in. (8 mm) × 0.118 in. (3 mm)
Synonym(e):
Spinbar® magnetic stirring fleas
About This Item
Empfohlene Produkte
Materialien
PTFE-coated
white
Leistungsmerkmale
ALNICO V coated magnet
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F37119-0083
L × Durchm.
0.315 in. (8 mm) × 0.118 in. (3 mm)
Verwandte Kategorien
Allgemeine Beschreibung
- Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
- Ideal for mixing of small volume samples in environmental testing and life science applications.
- Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
- Broad chemical resistance for use in a wide variety of applications.
- Autoclavable.
- PTFE-coated
- ALNICO V coated magnet
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BAF371190083-1EA | 4065269239024 |