BAF131900100
Clavies® Beutelklemmen
autoclave safe
Synonym(e):
bag seal
Alle Fotos(1)
About This Item
UNSPSC-Code:
24111500
NACRES:
NB.35
Empfohlene Produkte
Verpackung
pack of 100 ea
Hersteller/Markenname
Bel-Art F13190-0100
Verwandte Kategorien
Allgemeine Beschreibung
Open and close the clip with one hand. After autoclaving, clip holds the processed bag closed for final disposal. The closed internal dimension of the clip is 7/16 in., suitable for autoclave bags from 12 x 24 in. to 19 x 23 in. Do not use with larger bags.
- Holds filled autoclave bags closed during cycle
- Clip permits bag to safely vent internal pressure during autoclaving
Rechtliche Hinweise
Clavies is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)