Principaux documents
20627
Supelco
Septa GR-2
pkg of 1000 ea
About This Item
Code UNSPSC :
41115717
Conditionnement
pkg of 1000 ea
Diamètre
9.5 mm ( 3/8 in.)
Description générale
Température de l′injecteur : 50 °C à 200 °C
  • Septa en gomme de silicone gris peu coûteux, pour utilisation en routine et en isotherme.
Septum is a very important component of syringe injection system. They are usually fabricated from elastomeric, self-sealing materials such as silicone rubber, neoprene or fluoroelastomers. It is suitable to conveniently introducing the sample without causing the system to leak. Improper septa decomposes under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
Robert L. Grob, PhD, Eugene F. Barry, PhD
Modern Practice of Gas Chromatography, 472-472 (2004)
R.L. Grob, M.A. Kaiser
Environmental Problem Solving Using Gas and Liquid Chromatography, 166-166 (2000)