Principaux documents
Z744062
SealPlate® RollMate Dispenser with1 Roll of SealPlate® Film
case of 1 ea (500 Pre-Scored Sheets)
About This Item
Code UNSPSC :
41122410
Nomenclature NACRES :
NC.01
Conditionnement
case of 1 ea (500 Pre-Scored Sheets)
Fabricant/nom de marque
Excel Scientific SP2-RL-500
Paramètres
-40 °C temp. range (120 °C)
Technique(s)
ELISA: suitable
Description générale
  • An economical, efficient way to rapidly seal microplates; faster than applying single sheeted film
  • Self-standing, non-slip supports hold pre-scored film roll securely in place for quick and easy dispensing
  • Continuous backing liner can be torn off or fed directly into a waste basket for disposal
  • Roll tension can be easily adjusted to user preference (e.g. loose, tight)
  • SealPlate film minimizes evaporation, reduces cross-contamination and prevents spillage
  • 2.0 mil rigid polyester film features good clarity and easy handling
  • Eliminates the “edge effect”, excess evaporation in the outside wells, which can occur with other films and plate lids
  • Useful as a simple cover to protect samples from external contamination, dust and debris
  • High integrity packaging prevents damage during transit; poly bag eliminates dust and particulate contamination
Informations légales
RollMate is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.