Principaux documents
Z720984
Nunc® Cell Factory accessories
PTFE connector with O-ring, non-sterile, 2/cs
Synonyme(s) :
cell harvesting system, closed cell culture system
About This Item
Code UNSPSC :
41161502
Nomenclature NACRES :
NB.22
Stérilité
non-sterile
Conditionnement
pack of 1
case of 2
Fabricant/nom de marque
Nunc 173250
Autres remarques
Cell Factory accessories for filling and emptying the cell factories with media
Informations légales
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries