Principaux documents
Z331430
Lenz® round bottom flasks
capacity 250 mL, joint: ST/NS 14/23
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass 3.3
round bottom flask
single-neck
Joint
joint: ST/NS14/23
Agence
according to DIN 12 348
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Lenz 3.0014.49
Capacité
250 mL
Informations légales
Lenz is a registered trademark of Lenz GmbH