Principaux documents
TTRT96P250BW-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Semi-Skirted, PCR clean, volume 250 μL, blue plate (white wells), pack of 25
About This Item
Code UNSPSC :
41106300
Matériaux
blue plate (white wells)
polypropylene wells
Stérilité
non-sterile
Caractéristiques
skirt
PCR clean
Semi-Skirted
Conditionnement
pack of 25
Fabricant/nom de marque
Eppendorf® 951022043 (NA Only)
Technique(s)
PCR: suitable
Taille
96 well
Volume
250 μL
Volume de travail
250 μL
Description générale
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Application
Eppendorf twin.tec®Real-Time PCR Plates 96 is ideal for real time PCR and melting curve analysis.
Caractéristiques et avantages
  • White wells ensure better fluorescent light and other fluorescence assays reflection
  • For effective sealing rims are raised
  • Polypropylene wells ensure fast heat transfer and frames are made up of polycarbonate that provides easy and reliable handling
  • Extremely stable mechanically with skirted and semi-skirted frames
  • Reduced wall thickness ensures optimal heat transfer
  • Packed in Eppendorf® forensic DNA grade and lot-certified PCR clean for tested purity
  • With barcode available (upon request)
  • Autoclavable
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG