Principaux documents
OH30073977
Ohaus® Pen pH Meter
Model ST20C-C
About This Item
Code UNSPSC :
41115603
Matériaux
epoxy shaft
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30073977
Plage de mesure de la conductivité
0.00-199.9 μS/cm
Plage de mesure de la température
0.0-99.0 °C
Alimentation par piles
4 × 1.5 V, 200 hr life
Description générale
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Application
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Caractéristiques et avantages
Easily replaceable electrodes, automatic shut off feature
Informations légales
Ohaus is a registered trademark of Ohaus Corp.