Key Documents
MTCM5451
MTC Bio Triangle Spreader
L × W 30 mm × 207 mm, sterile, pack of 100 ea (10 x bags 10 ea)
Synonyme(s) :
cell spreader
About This Item
Code UNSPSC :
41122103
Nomenclature NACRES :
NB.17
Stérilité
sterile
Conditionnement
pack of 100 ea (10 x bags 10 ea)
Fabricant/nom de marque
MTC Bio M5451
L × l
30 mm × 207 mm
Description générale
Spreader Triangle, 30 x 207mm, 10 sterile peel bags of 10 spreaders, 100/pk
Caractéristiques et avantages
  • Spreads cultures and liquids evenly
  • Ideal for bacterial CFU counts
  • Smooth edges - minimizes gouging
  • Pre-sterilized - no flaming required
Informations légales
MTC is a trademark of MTC Bio