Principaux documents
EP5702705004
Eppendorf® Fixed-angle rotor
F-35-30-17, 10 x 15 ml, incl. 10 adapters Falcon tubes and 10 rubber mats
About This Item
Code UNSPSC :
41103909
Nomenclature NACRES :
NB.82
Caractéristiques
to hold 20 x 15 mL conical tubes
to hold 30 x 15 mL glass tubes
F-35-30-17, 10 x 15 ml, incl. 10 adapters Falcon tubes and 10 rubber mats
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 5702705004
Paramètres
4,400 max. RCF (2,750 xg)
Technique(s)
centrifugation: suitable
Compatibilité
for use with centrifuge models 5702/5702R/5702RH
Description générale
Fixed angle rotors for use with Eppendorf centrifuge models.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG