Principaux documents
EP0030625900
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
white polypropylene, V-bottom black wells, PCR clean, pkg of 240 ea
About This Item
Code UNSPSC :
41122107
Nomenclature NACRES :
NB.15
Matériaux
V-bottom black wells
white polypropylene
Conditionnement
pkg of 240 ea
Fabricant/nom de marque
Eppendorf® 0030625900
Technique(s)
PCR: suitable
Taille
384 wells
Description générale
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG