Principaux documents
CLS431328
Corning® CellBIND® Surface cell culture flasks
CellBIND 175cm2 angled neck cell culture flask w/ Vent Cap and Bar Code, PS, sterile, 84/cs
Synonyme(s) :
cell culture flasks, tissue culture flasks
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.13
Matériaux
angled neck
cap (vented)
high-density polyethylene cap
rectangular flask
rectangular polystyrene
Stérilité
sterile; γ-irradiated
Caractéristiques
graduations
Conditionnement
pack of 7
case of 84
Fabricant/nom de marque
Corning 431328
Volume maximum
250 mL
Superficie
175 cm2
Volume de travail
35-52.5 mL
Adéquation
suitable for (cell culture applications; optimal for adherent cell culturing)
Description générale
The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.
CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
  • Manufactured from optically clear virgin polystyrene.
  • Treated CellBIND surface treatment for enhanced cell attachment
  • 100% integrity tested.
  • Sterilized by gamma irradiation and certified nonpyrogenic
Informations légales
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.