CLS3839BC
Corning® 1536 well plate, with generic barcode
size 1536 wells, black/clear bottom polystyrene wells, low base, PDL, lid, sterile
Synonyme(s) :
1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, collagen coated plate
About This Item
Produits recommandés
Matériaux
black/clear bottom polystyrene wells
polystyrene plate
Stérilité
sterile
Caractéristiques
lid
PDL
low base
Conditionnement
pack of 10 ea
pack of 10
case of 50 ea
case of 50
Fabricant/nom de marque
Corning 3839BC
Volume maximum
12.5 μL
Taille
1536 wells
Volume de travail
8 μL
Adéquation
suitable for (cell cuture)
Catégories apparentées
Description générale
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
- Resistant to temperature ranges of -80ºC to +52ºC
- 2.8 mm distance between well bottoms to plate bottom
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
Microplate Features
- Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
- Superior flatness -minimizes variability within a plate and between plates
- Flat bottom wells are compatible with automated dispensers
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions.
- Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²)
- Working volume of up to 10 μL
Bar Code Details
Corning barcode labels have been quality tested for optimal readability, chemical resistance and temperature variation.
- Resistant to commonly used laboratory reagents (100% DMSO, Methanol, Ethanol, Ethyl Acetate, Methylene Chloride, Isopropanol, Acetronitrile, Ethanol/Water/TFA, Sodium Hydroxide, Hydrochloric Acid)
- Resistant to temperature ranges of -80ºC to +52ºC
Technical Details
- Bar Code labeling specifications
- 2.75" x 0.25" label size
- Code 128
- Code 3 of 9
- 10 mil narrow bar element (x-dimension = 0.010")
Global Trade Item Number
Référence | GTIN |
---|---|
CLS3839BC-50EA | 4061833884676 |
CLS3839BC-10EA | 4061833875698 |