Principaux documents
C4676
Sigma-Aldrich
Vertical mini-gel Comb
10 cm wide, for use with 1.0 mm x 14 wells
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Nom du produit
Combs for vertical mini-gel, 10 cm wide, size 1.0 mm × 14 wells
Taille
1.0 mm × 14 wells