Principaux documents
AXYT400LR
MAXYMum Recovery® ultra micro pipette tips
for Eppendorf® racked, size 0.5-10 μL, pkg of 5x960tips/cs
Synonyme(s) :
pipet tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Fabricant/nom de marque
Axygen Inc T-400-L-R
Corning
Conditionnement
pkg of 5x960tips/cs
Taille
0.5-10 μL
Volume
0.5—10 μL
Autres remarques
Browse all pipette tips
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.