Principaux documents
TZNFVSP20
Millipore
Steritest® Sampling Needle-Free Valve
sterile; ethylene oxide treated
About This Item
Code UNSPSC :
23151818
Nomenclature NACRES :
JA.12
Agence
USP 88
Niveau de qualité
Stérilité
sterile; ethylene oxide treated
Conditionnement
box of 10 units (2 valves per package)
Application(s)
pharmaceutical
sterility testing
Description générale
Steritest® Sampling Needle-Free Valve is designed as an accessory which is installed on Steritest® Neo Devices. The valve has a male luer slip and a female luer lock with a septum. The valve can be activated by a male luer without a needle.
The unique design allows aseptic installation (inside an isolator) post media filling step during sterility testing. The installation is recommended when a sampling of the media is planned.
Post installation, the media inside the canisters can be accessed through a luer syringe. Sampled media from Steritest® canisters can be further used for:
  • Turbidity detected during or after incubation
  • Use of rapid detection method (e.g: ATP measurement)
Application
Steritest® Sampling Needle-Free Valve is used to sample an aliquot of media from the canisters during or after incubation during sterility testing.
Informations légales
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany