PA18/111
NovAseptic® Valve, Pneumatic Plastic Actuator, Normally Closed, NC
About This Item
Code UNSPSC :
40141600
eCl@ss :
37051001
Produits recommandés
Matériaux
POM-C indicator
cover (Stainless Steel EN 1.4305/ASTM 303 and Plastic POM C)
Niveau de qualité
Gamme de produits
NovAseptic®
Paramètres
pressure range at -1--6 bar
temp. range at 0-70 °C
Poids
0.7 kg (1.5 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Autres remarques
Actuator, Pneumatic
Informations légales
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany